Learn More
PHRF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA564080
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: ASGRVQEAAR PEEVVSQTPL LRSRALVKRV TWNLQESESS APAEDRAPRA PLHRPQKPRE GAWDMEDVAP TGVRQAFSEL PF Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 64%.
Plays important roles in controlling the metabolism of fatty acids at differents levels. Acts as a magnesium-dependent phosphatidate phosphatase enzyme which catalyzes the conversion of phosphatidic acid to diacylglycerol during triglyceride, phosphatidylcholine and phosphatidylethanolamine biosynthesis in the reticulum endoplasmic membrane. Acts also as a nuclear transcriptional coactivator for PPARGC1A to modulate lipid metabolism.Specifications
PHRF1 | |
Polyclonal | |
Unconjugated | |
PHRF1 | |
AA673488; CTD-binding SR-like protein rA9; Kiaa1542; PHD and RING finger domain-containing protein 1; PHD and ring finger domains 1; phrf1; PPP1R125; protein phosphatase 1, regulatory subunit 125; RNF221 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57661 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P1Y6 | |
PHRF1 | |
Recombinant protein corresponding to Human PHRF1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only