Läs mer
Phthalate esters Mixture for GB/T 21294-2014 2000 μg/mL in Hexane, Dr. Ehrenstorfer
Produktkod. 30317070
Change view
Kvantitet:
1 mL
Produktkod. Kvantitet
30317070 1 mL
1 options
Produktkod. 30317070 Leverantör Dr. Ehrenstorfer Leverantörsnummer DREGA09000532HE
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dibutyl phthalate∣Diethyl phthalate∣Di-n-octyl phthalate∣Phthalic acid, benzylbutyl ester∣Phthalic acid, bis-2-ethylhexyl ester∣Phthalic acid, bis-methyl ester
CAS 84-74-2, 84-66-2, 117-84-0, 85-68-7, 117-81-7, 131-11-3
Kvantitet 1 mL
Koncentration eller sammansättning (efter analyt eller komponenter) Dibutyl phthalate 2000 μg/mL∣Diethyl phthalate 2000 μg/mL∣Di-n-octyl phthalate 2000 μg/mL∣Phthalic acid, benzylbutyl ester 2000 μg/mL∣Phthalic acid, bis-2-ethylhexyl ester 2000 μg/mL∣Phthalic acid, bis-methyl ester 2000 μg/mL