missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phthalic acid, bis-2-ethylhexyl ester, Dr. Ehrenstorfer
Specifications
Specifications
| Kemiskt namn eller material | Phthalic acid, bis-2-ethylhexyl ester |
| CAS | 117-81-7 |
| Analyt- eller komponentnamn | Phthalic acid, bis-2-ethylhexyl ester |
| Frakt skick | +20°C |
| Molekylformel | C24 H38 O4 |
| Certifikat och efterlevnad | ISO 17034 |
| Synonym | Diethylhexylphthalate, Bis-(2-ethylhexyl) Phthalate, Plastic additive 01, DEHP |
| InChI formel | InChI=1S/C24H38O4/c1-5-9-13-19(7-3)17-27-23(25)21-15-11-12-16-22(21)24(26)28-18-20(8-4)14-10-6-2/h11-12,15-16,19-20H,5-10,13-14,17-18H2,1-4H3 |
| Lösningstyp | Neat |
| LEDER | CCCCC(CC)COC(=O)c1ccccc1C(=O)OCC(CC)CCCC |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?