Läs mer
Phthalic Acid Bis(3,7-dimethyloctyl) Ester, TRC
Produktkod. 30428641
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30428641 100 mg
30428378 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30428641 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP384535100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 316808-86-3
Molekylformel C28H46O4
Molekylvikt (g/mol) 446.66
Synonym 1,2-Benzenedicarboxylic Acid 1,2-Bis(3,7-dimethyloctyl) Ester, 1,2-Benzenedicarboxylic Acid Bis(3,7-dimethyloctyl) Ester
IUPAC-namn bis(3,7-dimethyloctyl) phthalate
LEDER O=C(OCCC(C)CCCC(C)C)C=1C=CC=CC1C(=O)OCCC(C)CCCC(C)C
Specifikationer
Kemiskt namn eller material Phthalic Acid Bis(3,7-dimethyloctyl) Ester
Kvantitet 100 mg
InChI formel InChI=1S/C28H46O4/c1-21(2)11-9-13-23(5)17-19-31-27(29)25-15-7-8-16-26(25)28(30)32-20-18-24(6)14-10-12-22(3)4/h7-8,15-16,21-24H,9-14,17-20H2,1-6H3
Formel vikt 446.34