Läs mer
Phthalic acid, bis-isohexyl ester (mixture of isomers), Dr. Ehrenstorfer
Specifikationer
Specifikationer
| Kemiskt namn eller material | Phthalic acid, bis-iso-hexyl ester |
| CAS | 68515-50-4 |
| Molekylformel | C20H30O4 |
| Kvantitet | 100 mg |
| Synonym | Dihexylphthalate, Phthalic Acid, Bis-isohexyl Ester (Mixture of Isomers), Phthalic acid bis-iso-hexyl ester, 1,2-Benzenedicarboxylic acid, 1,2-dihexyl ester (branched and linear), 1,2-Benzenedicarboxylic acid, dihexyl ester, branched and linear |
| LEDER | CCOC(=O)c1c(cccc1)C(O)=O |
| Molekylvikt (g/mol) | 334.45 |
| Formel vikt | 334.21 g/mol |