Läs mer
Phthalic Acid Cyclohexyl Isobutyl Ester, TRC
Produktkod. 30420239
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30420239 50 mg
1 options
Produktkod. 30420239 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP38452050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Phthalic Acid Cyclohexyl Isobutyl Ester
CAS 5334-09-8
Molekylformel C18H24O4
Kvantitet 50 mg
Synonym NSC 2387, NSC 67410, 1,2-Benzenedicarboxylic Acid','Cyclohexyl 2-Methylpropyl Ester, 1,2-Benzenedicarboxylic Acid','1-Cyclohexyl 2-(2-Methylpropyl) Ester, Phthalic Acid Cyclohexyl Isobutyl Ester
InChI formel InChI=1S/C18H24O4/c1-13(2)12-21-17(19)15-10-6-7-11-16(15)18(20)22-14-8-4-3-5-9-14/h6-7,10-11,13-14H,3-5,8-9,12H2,1-2H3
LEDER O=C(OCC(C)C)C=1C=CC=CC1C(=O)OC2CCCCC2
IUPAC-namn 2-O-cyclohexyl 1-O-(2-methylpropyl) benzene-1,2-dicarboxylate
Molekylvikt (g/mol) 304.38
Formel vikt 304.1675