missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PHYH (Human) Recombinant Protein
Human PHYH full-length ORF ( AAH29512, 1 a.a. - 338 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005264-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MEQLRAAARLQIVLGHLGRPSAGAVVAHPTSGTISSASFHPQQFQYTLDNNVLTLEQRKFYEENGFLVIKNLVPDADIQRFRNEFEKICRKEVKPLGLTVMRDVTISKSEYAPSEKMITKVQDFQEDKELFRYCTLPEILKYVECFTGPNIMAMHTMLINKPPDSGKKTSRHPLHQDLHYFPFRPSDLIVCAWTAMEHISRNNGCLVVLPGTHKGSLKPHDYPKWEGGVNKMFHGIQDYEENKARVHLVMEKGDTVFFHPLLIHGSGQNKTQGFRKAISCHFASADCHYIDVKGTSQENIEKEVVGIAHKFFGAENSVNLKDIWMFRARLVKGERTNL
Specifications
AAH29512 | |
phytanoyl-CoA 2-hydroxylase | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PHYH | |
GST |
5264 | |
Wheat germ expression system | |
25 μg | |
LN1, LNAP1, PAHX, PHYH1, RD | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |