missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHYHIPL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | PHYHIPL |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16836724
|
Proteintech
16937-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16826724
|
Proteintech
16937-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
16937-1-AP detects 39 kDa and 42-45 kDa isoforms in SDS-PAGE.
PHYHIPL may play a role in the development of the central system.Specifications
PHYHIPL | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
Q6AYN4, Q8BGT8, Q96FC7 | |
309901, 70911, 84457 | |
PHYHIPL Fusion Protein Ag10435 | |
Primary | |
-20°C | |
PHYHIPL |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
KIAA1796, PHYHIPL | |
Phyhipl | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |