missing translation for 'onlineSavingsMsg'
Learn More

Phytane, TRC

Product Code. 30421665
Change view
Click to view available options
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30421665 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30421665 Supplier Toronto Research Chemicals Supplier No. TRCP39900525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Phytane
CAS 638-36-8
Molekylformel C20 H42
Kvantitet 25 mg
Synonym Hexadecane','2,6,10,14-tetramethyl-, Phytane (6CI), 2,6,10,14-Tetramethylhexadecane, Phytan, Tetrahydroneophytadiene
InChI formel InChI=1S/C20H42/c1-7-18(4)12-9-14-20(6)16-10-15-19(5)13-8-11-17(2)3/h17-20H,7-16H2,1-6H3
LEDER CCC(C)CCCC(C)CCCC(C)CCCC(C)C
IUPAC-namn 2,6,10,14-tetramethylhexadecane
Molekylvikt (g/mol) 282.55
Formel vikt 282.3287
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.