Läs mer
Phytanoic Acid, TRC
Produktkod. 30413508
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30413508 1 g
30418093 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30413508 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP1472001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Phytanoic Acid
CAS 14721-66-5
Anteckningar om renhetsgrad HPLC
Molekylformel C20 H40 O2
Kvantitet 1 g
Synonym 3,7,11,15-Tetramethylhexadecanoic Acid, NSC 108698, Phytanic Acid
InChI formel InChI=1S/C20H40O2/c1-16(2)9-6-10-17(3)11-7-12-18(4)13-8-14-19(5)15-20(21)22/h16-19H,6-15H2,1-5H3,(H,21,22)
LEDER CC(C)CCCC(C)CCCC(C)CCCC(C)CC(=O)O
IUPAC-namn 3,7,11,15-tetramethylhexadecanoic acid
Molekylvikt (g/mol) 312.53
Formel vikt 312.3028