Läs mer
Phytic Acid Dodecasodium Salt Hydrate, TRC
Produktkod. 30408948
Change view
Kvantitet:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30408948 1 g
30414061 5 g
30414060 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30408948 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP3987131G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 123408-98-0
Molekylformel C6H6Na12O24P6 . xH2O
Molekylvikt (g/mol) 941.83
Synonym Hexakis(Dihydrogen Phosphate) myo-Inositol Dodecasodium Salt Hydrate
IUPAC-namn dodecasodium;(2,3,4,5,6-pentaphosphonatooxycyclohexyl) phosphate;hydrate
LEDER O.[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[O-]P(=O)([O-])O[C@@H]1[C@H](OP(=O)([O-])[O-])[C@@H](OP(=O)([O-])[O-])[C@H](OP(=O)([O-])[O-])[C@@H](OP(=O)([O-])[O-])[C@@H]1OP(=O)([O-])[O-]
Specifikationer
Kemiskt namn eller material Phytic Acid Dodecasodium Salt Hydrate
Kvantitet 1 g
InChI formel InChI=1S/C6H18O24P6.12Na.H2O/c7-31(8,9)25-1-2(26-32(10,11)12)4(28-34(16,17)18)6(30-36(22,23)24)5(29-35(19,20)21)3(1)27-33(13,14)15;;;;;;;;;;;;;/h1-6H,(H2,7,8,9)(H2,10,11,12)(H2,13,14,15)(H2,16,17,18)(H2,19,20,21)(H2,22,23,24);;;;;;;;;;;;;1H2/q;12*+1;/p-12/t1-,2-,3-,4+,5-,6-;;;;;;;;;;;;;
Formel vikt 941.6553