Läs mer
Phytic Acid (Technical Grade), TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Phytic Acid |
| CAS | 83-86-3 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C6 H18 O24 P6 |
| Kvantitet | 250 mg |
| Synonym | myo-Inositol','1,2,3,4,5,6-hexakis(dihydrogen phosphate), Inositol','hexakis(dihydrogen phosphate)','myo- (8CI), myo-Inositol','hexakis(dihydrogen phosphate) (9CI), Alkalovert, Dermofeel PA 3, Exfoderm, Exfoderm Forte, Fytic acid, IP6, Inositol 1,2,3,4,5,6-hexakisphosphate, Inositol hexakis(phosphate), Inositol hexaphosphate, InsP6, Phyliance, Phytic acid, RP 3000, RP 3000 (rust inhibitor), WHL, WHL (depressant), meso-Inositol hexaphosphate, myo-Inositol hexakis(phosphate), myo-Inositol hexaphosphate |
| InChI formel | InChI=1S/C6H18O24P6/c7-31(8,9)25-1-2(26-32(10,11)12)4(28-34(16,17)18)6(30-36(22,23)24)5(29-35(19,20)21)3(1)27-33(13,14)15/h1-6H,(H2,7,8,9)(H2,10,11,12)(H2,13,14,15)(H2,16,17,18)(H2,19,20,21)(H2,22,23,24)/t1-,2-,3-,4+,5-,6- |
| LEDER | OP(=O)(O)O[C@@H]1[C@@H](OP(=O)(O)O)[C@H](OP(=O)(O)O)[C@H](OP(=O)(O)O)[C@H](OP(=O)(O)O)[C@H]1OP(=O)(O)O |
| IUPAC-namn | (2,3,4,5,6-pentaphosphonooxycyclohexyl) dihydrogen phosphate |
| Molekylvikt (g/mol) | 660.04 |
| Visa mer |