Läs mer
Phytonadione trans Epoxy (contains up to 20% cis isomer), TRC
Produktkod. 30135477
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135477 100 mg
30132586 50 mg
30005041 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135477 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP123530100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 52691-91-5
Molekylformel C31 H46 O3
Molekylvikt (g/mol) 466.7
Synonym Phytonadione trans Epoxy (contains up to 20% cis isomer);Phylloquinone Epoxide, 2-Methyl-3-((3-methyl-3-(4,8,12-trimethyltridecyl)oxiran-2-yl)methyl)naphthalene-1,4-dione
IUPAC-namn 2-methyl-3-[[3-methyl-3-[(4S,8S)-4,8,12-trimethyltridecyl]oxiran-2-yl]methyl]naphthalene-1,4-dione
LEDER CC(C)CCC[C@H](C)CCC[C@H](C)CCCC1(C)OC1CC2=C(C)C(=O)c3ccccc3C2=O
Specifikationer
Kemiskt namn eller material Phytonadione trans Epoxy
Kvantitet 100 mg
InChI formel InChI=1S/C31H46O3/c1-21(2)12-9-13-22(3)14-10-15-23(4)16-11-19-31(6)28(34-31)20-27-24(5)29(32)25-17-7-8-18-26(25)30(27)33/h7-8,17-18,21-23,28H,9-16,19-20H2,1-6H3/t22-,23-,28?,31?/m0/s1
Formel vikt 466.3447