Läs mer
(+)-Phytone, TRC
Produktkod. 30420853
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30420853 100 mg
1 options
Produktkod. 30420853 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP398050100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (+)-Phytone
CAS 16825-16-4
Molekylformel C18H36O
Kvantitet 100 mg
Synonym (6R,10R)-6,10,14-Trimethylpentadecan-2-one, (R,R)-Phytone, Phytol Ketone, Phytone, Phytone (ketone)
InChI formel InChI=1S/C18H36O/c1-15(2)9-6-10-16(3)11-7-12-17(4)13-8-14-18(5)19/h15-17H,6-14H2,1-5H3/t16-,17-/m1/s1
LEDER O=C(C)CCC[C@H](C)CCC[C@H](C)CCCC(C)C
Molekylvikt (g/mol) 268.48
Formel vikt 268.2766