Learn More
PIAS3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552288
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PTKKHCSVTS AAIPALPGSK GVLTSGHQPS SVLRSPAMGT LGGDFLSSLP LHEYPPAFPL GADIQGLDLF SFLQTESQHY GPSVITSLDE QDALGHFFQY RGTPSHFLGP LAPTLGSSHC SATPAPPPGR VSSIVAPG Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 93%.
The PIAS (protein inhibitor of activated STAT) proteins play a crucial role as transcriptional coregulators in various cellular pathways, including the STAT, p53 and the steroid hormone signaling pathway. The PIAS protein family includes at least five evolutionarily conserved genes, including PIAS3. The major function of the PIAS proteins is the control of gene transcription and can also act as small ubiquitin-like-modifier (SUMO) E3 ligases. PIAS3 binds specifically to STAT3 following the stimulation of STAT3. Increased expression of PIAS3 has been observed in several human cancers, including lung, breast, and brain tumors, but not in anaplastic lymphoma kinase-positive T/null-cell lymphomas, indicating that PIAS3 plays multiple roles in different tissue and cell types.Specifications
PIAS3 | |
Polyclonal | |
Unconjugated | |
PIAS3 | |
E3 SUMO-protein ligase PIAS3; E3 SUMO-protein transferase PIAS3; KChAP; PIAS3; Pias3l; potassium channel regulatory protein KChAP; potassium channel-associated protein; protein inhibitor of activated STAT 3; protein inhibitor of activated STAT protein 3; protein inhibitor of activated STAT, 3; zinc finger, MIZ-type containing 5; ZMIZ5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10401 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y6X2 | |
PIAS3 | |
Recombinant protein corresponding to Human PIAS3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only