Läs mer
Picloram, TRC
Produktkod. 30420238
Change view
Kvantitet:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30420238 1 g
30410217 10 g
30416616 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30420238 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP4367001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1918-02-1
Molekylformel C6 H3 Cl3 N2 O2
Molekylvikt (g/mol) 241.46
Synonym 2-Pyridinecarboxylic acid, 4-amino-3,5,6-trichloro-, Picolinic acid, 4-amino-3,5,6-trichloro- (8CI), 4-Amino-3,5,6-trichloro-2-pyridinecarboxylic acid, 3,5,6-Trichloro-4-aminopicolinic acid, 4-Amino-3,5,6-trichloro-2-picolinic acid, 4-Amino-3,5,6-trichloropicolinic acid, 4-Amino-3,5,6-trichloropyridine-2-carboxylic acid, 4-Aminotrichloropicolinic acid, ATCP, Grazon PC, NSC 233899, Padron, Padron (herbicide), Picloram, Tordon
IUPAC-namn 4-amino-3,5,6-trichloropyridine-2-carboxylic acid
LEDER Nc1c(Cl)c(Cl)nc(C(=O)O)c1Cl
Specifikationer
Kemiskt namn eller material Picloram
Kvantitet 1 g
InChI formel InChI=1S/C6H3Cl3N2O2/c7-1-3(10)2(8)5(9)11-4(1)6(12)13/h(H2,10,11)(H,12,13)
Formel vikt 239.926