Läs mer
Picloxydine Digluconate, LoGiCal
Produktkod. 30458026
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30458026 10 mg
1 options
Produktkod. 30458026 Leverantör LoGiCal Leverantörsnummer LGCFOR3194.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Picloxydine Digluconate
CAS 29147-50-0
Analyt- eller komponentnamn Picloxydine Digluconate
Frakt skick Room Temperature
Molekylformel C20 H24 Cl2 N10 . 2 C6 H12 O7
Certifikat och efterlevnad ISO 9001
Synonym D-Gluconic acid, compd. with N,N''-bis[[(4-chlorophenyl)amino]iminomethyl]-1,4-piperazinedicarboximidamide (2:1) (9CI), Gluconic acid, compd. with N,N''-bis[(p-chlorophenyl)amidino]-1,4-piperazinedicarboxamidine (2:1), D- (8CI), 1,4-Piperazinedicarboxamidine, N,N''-bis[(p-chlorophenyl)amidino]-, di-D-gluconate (8CI), 1,4-Piperazinedicarboximidamide, N,N''-bis[[(4-chlorophenyl)amino]iminomethyl]-, di-D-gluconate (9CI), Picloxydine digluconate
InChI formel InChI=1S/C20H24Cl2N10.2C6H12O7/c21-13-1-5-15(6-2-13)27-17(23)29-19(25)31-9-11-32(12-10-31)20(26)30-18(24)28-16-7-3-14(22)4-8-16;2*7-1-2(8)3(9)4(10)5(11)6(12)13/h1-8H,9-12H2,(H4,23,25,27,29)(H4,24,26,28,30);2*2-5,7-11H,1H2,(H,12,13)/t;2*2-,3-,4+,5-/m.11/s1
Lösningstyp Neat
LEDER OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(=O)O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(=O)O.Clc1ccc(NC(=N)NC(=N)N2CCN(CC2)C(=N)NC(=N)Nc3ccc(Cl)cc3)cc1
IUPAC-namn 1-N,4-N-bis[N-(4-chlorophenyl)carbamimidoyl]piperazine-1,4-dicarboximidamide;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid
Industry Type Clinical Testing
Molekylvikt (g/mol) 867.69
Kvantitet 10 mg
Formel vikt 866.2729 g/mol