Learn More
Ansell Picosoft DG Light Duty Polyamide Gloves with PVC-Dotted Palm
Click to view available options
Size:
6
7
8
9
10
11
Description
- Enhanced grip capabilities with dipped fingertips and a PVC-dotted palm
- Ideal for light and delicate applications in dry environments
- Ensures tactility and dexterity for precise and safe handling
Specifications
Certifications/Compliance (Europe) | EN 420:2003 + A1:2009, REACH Compliant |
Coating Material | PVC Dots |
Color | Gray |
Hand | Specific |
Material | Polyamide |
Size | 6 |
EN388 Standard | 2121A |
Glove Cuff Style | Knit |
Series | Picosoft DG |