missing translation for 'onlineSavingsMsg'
Learn More

Ansell Picostar C366 Light Duty Nylon Gloves with PVC-Dotted Palm

Catalog Number 16885330
Click to view available options
Size:
7
8
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Low lint gloves are ideal for lightweight applications and feature PVC dots on the palm to enhance grip capabilities.

  • PVC dots on the palm enhance grip capabilities
  • High degree of comfort, flexibility, and dexterity
  • Low lint for cleanroom applications
  • Resistant to abrasions
Description Low Lint
Coating Material PVC Dots
Color White
Hand Specific
Material Nylon
Size 10
EN388 Standard 2141A
For Use With (Application) Automotive Assembly, General Handling, and Parts Assembly
Glove Cuff Style Knit
Series C366
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'