Learn More
Thermo Scientific™ Pierce™ Disposable Plastic Columns
Empty drip columns (0.5 to 5 mL) with matching frits and caps for packing beaded affinity or desalting resins to use in gravity-flow purification techniques.
786.00 SEK - 2640.00 SEK
Products 5
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10066073 View Documents Thermo Scientific™
29920
Disposable Columns, 2mL 2335.00 SEK
Pack of 100 View alternative products
Save Up to 
»
EU Stock 17
Add to basket
10331064 View Documents Thermo Scientific™
29922
Disposable Columns, 5mL 2155.00 SEK
Pack of 100 View alternative products
Save Up to 
»
EU Stock 14
Add to basket
11825845 View Documents Thermo Scientific™
29923
Disposable Column Funnels 1328.00 SEK
Pack of 50 View alternative products
Save Up to 
»
EU Stock 7
Add to basket
10753485 View Documents Thermo Scientific™
29924
Disposable Columns, 10mL 2640.00 SEK
Pack of 100 View alternative products
Save Up to 
»
EU Stock 25
Add to basket
10763485 View Documents Thermo Scientific™
29925
Disposable Column Trial Pack 786.00 SEK
Each View alternative products
Save Up to 
»
EU Stock 7
Add to basket
Description
Description
These bulk packages of empty columns are economical and convenient devices for packing affinity purification supports and desalting resins for gravity-flow procedures. Three columns sizes are offered, suitable for procedures requiring 0.5 to 10mL resin-bed volumes. The plastic columns are supplied with matching pairs of polyethylene discs and polypropylene caps (top and bottom). The columns are economically-priced for single use and sufficiently durable to prepare and store columns for multiple uses.
Highlights:
  • For bench top gravity-flow affinity purification and size exclusion chromatography
  • Supplied complete with porous polyethylene discs, stoppers and end caps
  • Columns can be pre-packed and stored until needed
  • Compatible with most types of aqueous buffer eluents commonly used in chromatography
  • Plastic funnels can be attached to expand the buffer reservoir for increased flow rates
Includes:
Trial pack contains two columns of each available size (six columns total), matching column accessories (discs and caps), and one funnel (column reservoir extender)