Learn More
Thermo Scientific™ DST (disuccinimidyl tartrate)
Crosslink amines to amines with disuccinimidyl tartrate, composed of NHS-ester groups and a diol-containing spacer arm (6.4A) that is periodate-cleavable.
Brand: Thermo Scientific™ 20589
Additional Details : CAS Number : 62069-75-4 Weight : 0.09000kg
Description
Thermo Scientific™ Pierce DST, also called disuccinimidyl tartrate, is a water-soluble crosslinker that contains amine-reactive NHS ester ends around a 4-atom spacer arm, whose central diol can be cleaved by periodate oxidation.
Highlights:
- Cleavable by Sodium meta-Periodate
- Ideal for applications in which crosslink cleavability is desired without disturbing protein S-S bonds
- DST is soluble in water, DMF or DMSO at a concentration of ∽10mM
- Reactive groups: NHS ester (homobifunctional)
- Reactive toward: amino groups
Specifications
Amine-Amine | |
DST | |
No | |
Upon receipt store desiccated at 4°C. | |
Ambient | |
Chemical Labeling | |
NHS Ester | |
Powder | |
Water | |
Yes |
By Periodate | |
344.23 | |
6.4 Å | |
Yes | |
Pierce™ | |
Homobifunctional | |
Short (<10 Å) | |
50 mg | |
Standard | |
Crosslinker |
For Research Use Only. Not for use in diagnostic procedures.