Learn More
Thermo Scientific™ Pierce™ High Binding Streptavidin Coated Plates
Bind biotinylated oligonucleotides and peptides with higher binding efficiency than other commercially available streptavidin-coated plates.
1756.00 SEK - 3640.00 SEK
Specifications
Binding Target Biotin
Quantity 5 plates
View More Specs
Products 7
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10752024 View Documents Thermo Scientific™
15500
Pierce Streptavidin Coated High Capacity Plates, Clear, 96-Well 1756.00 SEK
Pack of 5 View alternative products
Save Up to 
»
EU Stock 68
Add to basket
10666234 View Documents Thermo Scientific™
15501
Pierce Streptavidin Coated High Capacity Plates, Clear, 96-Well Strip 2015.00 SEK
Pack of 5 View alternative products
Save Up to 
»
EU Stock 27
Add to basket
10003413 View Documents Thermo Scientific™
15502
Pierce Streptavidin Coated High Capacity Plates, White, 96-Well 1966.00 SEK
Pack of 5 View alternative products
Save Up to 
»
EU Stock 7
Add to basket
10532964 View Documents Thermo Scientific™
15503
Pierce Streptavidin Coated High Capacity Plates, Black, 96-Well 1966.00 SEK
Pack of 5 View alternative products
Save Up to 
»
EU Stock 8
Add to basket
10352444 View Documents Thermo Scientific™
15504
Pierce Streptavidin Coated High Capacity Plates, Clear, 384-Well 3640.00 SEK
Pack of 5 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 09-09-2019
Add to basket
10693954 View Documents Thermo Scientific™
15505
Pierce Streptavidin Coated High Capacity Plates, White, 384-Well 3635.00 SEK
Pack of 5 View alternative products
Save Up to 
»
EU Stock 8
Add to basket
10097683 View Documents Thermo Scientific™
15506
Pierce Streptavidin Coated High Capacity Plates, Black, 384-Well 3640.00 SEK
Pack of 5 View alternative products
Save Up to 
»
EU Stock 10
Add to basket
Description
Description
Pierce Streptavidin High Binding Capacity (HBC) Plates are prepared with an exclusive coating method that yields a streptavidin-coated plate with four to five times the binding capacity of other commercially available polystyrene assay plates. Using a Pierce Streptavidin HBC Plate can result in an assay with a broader dynamic range and better linearity, leading to improved assay precision. The plates are offered in several plate formats, plate colors and coating volumes.
Highlights:
  • Exclusive plate-coating technology – four to five times the capacity of typical biotin-binding plates
  • Broader dynamic range – extends the quantitative range so there is no need for dilutions
  • Better sensitivity – increased binding capacity allows direct detection of small ligands not observed with regular binding capacity plates
  • Superior assay precision – standard curve demonstrates greater linearity
  • Save time – pre-blocked to reduce number of assay steps
  • Flexible assay formats – offered in 96- and 384-well formats and in different colors
Specifications
Specifications
Biotin
5 plates