Learn More
Thermo Scientific™ Pierce™ Immobilized Reductant Column, 2 mL
Efficiently reduce protein or peptide disulfide bonds with this reusable column on with an immobilized thiol-based reducing agent for solid-phase disulfide bond reduction.
Brand: Thermo Scientific™ 77701
Additional Details : Weight : 0.80000kg
Description
Thermo Scientific™ Pierce Immobilized Reductant Column contains beaded resin to which an thiol-based reducing agent has been immobilized to enable solid-phase reduction of peptide and protein disulfide bonds.
Both the mercaptan and the DTT methods of reduction use soluble reducing agents. This requires that the reducing reagents be removed from the sample before further procedures can be performed. Measurements of free sulfhydryl content cannot be made in the presence of these agents, and they must be removed prior to crosslinking. Removal of a soluble reductant can be difficult when a peptide is involved, and extensive desalting may result in protein loss or extreme dilution when small quantities of protein are reduced. Reduce-Imm™ Reducing Columns were developed to eliminate these difficulties, and are highly effective for reducing disulfide bonds.Highlights:
- Eliminates soluble components which interfere with the assay of free thiol groups
- No need to remove reducing agent after the reaction
- Reduced protein, free of contaminating thiols, can be easily assayed using the BCA method
- Solid phase; protocols for both peptides and proteins
- Excellent protein recovery
- Regenerable matrix usable up to 4 times
Specifications
Upon receipt store at 4°C. | |
Liquid | |
Reductant Column |
Pierce™ | |
1 x 2 mL |
For Research Use Only. Not for use in diagnostic procedures.