Learn More
Thermo Scientific™ Sulfo-EMCS (N-ε-maleimidocaproyl-oxysulfosuccinimide ester)
Crosslink amines to sulfhydryls with this mid-length crosslinker composed of sulfo-NHS-ester and maleimide groups at ends of a water-soluble, 8-atom (9.4A) spacer arm.
Brand: Thermo Scientific™ 22307
Additional Details : Weight : 0.80000kg
Description
Thermo Scientific™ Pierce Sulfo-EMCS is a water-soluble amine-to-sulfhydryl crosslinker that contains NHS-ester and maleimide reactive groups at opposite ends of a medium-length spacer arm (9.4 Ås).
Highlights:
- Water-soluble
- Cleavable at pH 8.5 using hydroxylamine and heat
- Lactose dehydrogenase retained 60% of its activity after reversible crosslinking
- Reactive groups: sulfo-NHS esters (homobifunctional)
- Reactive toward: amino groups
Specifications
Amine-Sulfhydryl | |
Sulfo-EMCS | |
No | |
Upon receipt store desiccated at 4°C. | |
Ambient | |
Chemical Labeling | |
Maleimide, Sulfo-NHS Ester | |
Powder | |
Water | |
Yes |
No | |
410.33 | |
9.4 Å | |
No | |
Pierce™ | |
Heterobifunctional | |
Short (<10 Å) | |
50 mg | |
Standard | |
Crosslinker |
For Research Use Only. Not for use in diagnostic procedures.