Läs mer
STEMCELL Technologies Pifithrin-mu
Beskrivning
- Pifithrin-mu (PFT-μ) is an inhibitor of p53-mediated apoptosis, preventing p53 binding to Bcl-xL and Bcl-2 at the mitochondrial surface, without affecting p53 transactivational activities (Strom et al.) In vitro, PFT-μ binds both p53 (Kd = 0.82 mM) and Bcl-xL (Kd = 0.80 mM; Hagn et al.)
- In combination with Rho-associated coiled-coil containing protein kinase (ROCK) inhibitor Y-27632, improves cell recovery after cryopreservation (Xu et al.)
- Inhibits DNA damage-induced apoptosis in human embryonic stem cells (Qin et al.)
Specifikationer
Specifikationer
| CAS | 64984-31-2 |
| Synonym | 2-Phenylethynesulfonamide; PFT-μ; Pifithrin-μ |
| Kvantitet | 10 mg |
| Molekylformel | C8H7NO2S |
| Renhet | ≥98% |