Läs mer
Pigment Red 146 (Technical Grade), TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Pigment Red 146 (Technical Grade) |
| CAS | 5280-68-2 |
| Molekylformel | C33H27ClN4O6 |
| Kvantitet | 10 g |
| Synonym | Renol Carmine FBB-H, Sandye Super Pink FBL, Sanyo Pigment Pink 4602, Seikafast Carmine 3870, Sunbrite Red 146, Symuler Fast Red 4195, Symuler Fast Red 4580, Symuler Fast Red 4590, Vulcan Fast Carmine FBB, N-(4-Chloro-2,5-dimethoxyphenyl)-3-hydroxy-4-[2-[2-methoxy-5-[(phenylamino)carbonyl]phenyl]diazenyl]-2-naphthalenecarboxamide, C.I. 12485, C.I. PR 146, C.I. Pigment Red 146, Fuji Fast Carmine 550, Helio Fast Carmine BB, Hydrocolor Red, Karmine FBB 02, Lionol Red 5620, Microlith Red 3R-A, Naphtol Carmine 4635C, PR 146, Permanent Carmine FBB, Permanent Carmine FBB 02, Permanent Carmine FBB 02JP, Permanent Carmine FBB Extra, Pigment Pink 4602 |
| InChI formel | InChI=1S/C33H27ClN4O6/c1-42-27-14-13-20(32(40)35-21-10-5-4-6-11-21)16-26(27)37-38-30-22-12-8-7-9-19(22)15-23(31(30)39)33(41)36-25-18-28(43-2)24(34)17-29(25)44-3/h4-18,39H,1-3H3,(H,35,40)(H,36,41) |
| LEDER | COc1cc(NC(=O)c2cc3ccccc3c(N=Nc4cc(ccc4OC)C(=O)Nc5ccccc5)c2O)c(OC)cc1Cl |
| IUPAC-namn | N-(4-chloro-2,5-dimethoxyphenyl)-3-hydroxy-4-[(E)-[2-methoxy-5-(phenylcarbamoyl)phenyl]diazenyl]naphthalene-2-carboxamide |
| Molekylvikt (g/mol) | 611.04 |
| Formel vikt | 610.1619 |