Läs mer
Pigment Red 63:1 (Technical Grade), TRC
Produktkod. 30425360
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30425360 100 mg
30408907 50 mg
30423677 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30425360 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP437773100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6417-83-0
Molekylformel C21H12CaN2O6S
Molekylvikt (g/mol) 460.5
Synonym 2-Naphthalenecarboxylic acid, 3-hydroxy-4-[2-(1-sulfo-2-naphthalenyl)diazenyl]-, calcium salt (1:1) (ACI), 2-Naphthalenecarboxylic acid, 3-hydroxy-4-[(1-sulfo-2-naphthalenyl)azo]-, calcium salt (1:1) (9CI), C.I. Pigment Red 63, calcium salt (1:1) (8CI), 1952 Red, Bordeaux 10B, Bordeaux Toner BFR, C.I. 15880:1, C.I. Pigment Red 63:1, D and C Red 34 Calcium Lake, D and C Red No. 34, D&C Red 34, D&C Red 34 Calcium Lake, D&C Red 34, Deep Maroon Lake, Eljon Claret YS, Fanchon Maroon, HK Formula K 7122, Japan Red 220, Japan Red No. 220, Lake Bordeaux BC, Lithol Bordeaux 4761, Lithol Maroon ML 1320, Pigment Red 63:1, Red 220, Red 34 Lake, Red No. 220, 3-Hydroxy-4-[(1-sulfo-2-naphthalenyl)azo]-2-naphthalenecarboxylic acid calcium salt, Calcium 3-hydroxy-4-[(1-sulfonato-2-naphthyl)azo]-2-naphthoate, 3-Hydroxy-4-[2-(1-sulfo-2-naphthalenyl)diazenyl]-2-naphthalenecarboxylic acid calcium salt
IUPAC-namn calcium;3-hydroxy-4-[(1-sulfonatonaphthalen-2-yl)diazenyl]naphthalene-2-carboxylate
LEDER [Ca+2].Oc1c(cc2ccccc2c1N=Nc3ccc4ccccc4c3S(=O)(=O)[O-])C(=O)[O-]
Specifikationer
Kemiskt namn eller material Pigment Red 63:1 (Technical Grade)
Kvantitet 100 mg
InChI formel InChI=1S/C21H14N2O6S.Ca/c24-19-16(21(25)26)11-13-6-2-3-7-14(13)18(19)23-22-17-10-9-12-5-1-4-8-15(12)20(17)30(27,28)29;/h1-11,24H,(H,25,26)(H,27,28,29);/q;+2/p-2
Formel vikt 460.0042