Läs mer
Pigment Yellow 14 (Technical Grade), TRC
Produktkod. 30412041
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30412041 100 mg
1 options
Produktkod. 30412041 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP437843100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Pigment Yellow 14
CAS 5468-75-7
Molekylformel C34 H30 Cl2 N6 O4
Kvantitet 100 mg
Synonym Benzidine Yellow OT (6CI), Butanamide','2,2'-[(3,3'-dichloro[1,1'-biphenyl]-4,4'-diyl)bis(azo)]bis[N-(2-methylphenyl)-3-oxo- (9CI), C.I. Pigment Yellow 14 (7CI,8CI), 2,2'-[(3,3'-Dichloro[1,1'-biphenyl]-4,4'-diyl)bis(2,1-diazenediyl)]bis[N-(2-methylphenyl)-
InChI formel InChI=1S/C34H30Cl2N6O4/c1-19-9-5-7-11-27(19)37-33(45)31(21(3)43)41-39-29-15-13-23(17-25(29)35)24-14-16-30(26(36)18-24)40-42-32(22(4)44)34(46)38-28-12-8-6-10-20(28)2/h5-18,31-32H,1-4H3,(H,37,45)(H,38,46)
LEDER CC(=O)C(N=Nc1ccc(cc1Cl)c2ccc(N=NC(C(=O)C)C(=O)Nc3ccccc3C)c(Cl)c2)C(=O)Nc4ccccc4C
IUPAC-namn 2-[(E)-[2-chloro-4-[3-chloro-4-[(E)-[1-(2-methylanilino)-1,3-dioxobutan-2-yl]diazenyl]phenyl]phenyl]diazenyl]-N-(2-methylphenyl)-3-oxobutanamide
Molekylvikt (g/mol) 657.546
Formel vikt 656.171