Läs mer
Pigment Yellow 180, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Pigment Yellow 180 |
| CAS | 77804-81-0 |
| Molekylformel | C36H32N10O8 |
| Kvantitet | 1 g |
| Synonym | 2,2'-[1,2-Ethanediylbis(oxy-2,1-phenyleneazo)]bis[N-(2,3-dihydro-2-oxo-1H-benzimidazol-5-yl)-3-oxo-butanamide, BY 2000GT, C.I. 21290, C.I. PY 180, C.I. Pigment Yellow 180, CFY 313-2, Chromofine Yellow 313-2, Chromofine Yellow 6280JC, Diaplast Fast Yellow HG, Fast Yellow HG, Hansa Brilliant Yellow 5GYC4, Hostaperm Yellow HG, Liojet Yellow, Novoperm P-HG, Novoperm Yellow P-H 9, Novoperm Yellow P-HG, Novoperm Yellow P-HGVP 2066, PV Fast Yellow HG, PY 180, Pigment Yellow 180, Pigment Yellow JH-GT, Symuler Fast Yellow BY 2000GT, Toner Y HG, Toner Yellow HG, Toner Yellow HG-VP 2155, Yellow HG-AF, Yellow HG-AF LP 901, Yellow HG-AF-LP 901 |
| InChI formel | InChI=1S/C36H32N10O8/c1-19(47)31(33(49)37-21-11-13-23-27(17-21)41-35(51)39-23)45-43-25-7-3-5-9-29(25)53-15-16-54-30-10-6-4-8-26(30)44-46-32(20(2)48)34(50)38-22-12-14-24-28(18-22)42-36(52)40-24/h3-14,17-18,31-32H,15-16H2,1-2H3,(H,37,49)(H,38,50)(H2,39,41,51)(H2,40,42,52) |
| LEDER | O=C1NC2=C(C=C(NC(C(C(C)=O)/N=N/C3=CC=CC=C3OCCOC4=C(/N=N/C(C(NC5=CC=C(NC(N6)=O)C6=C5)=O)C(C)=O)C=CC=C4)=O)C=C2)N1 |
| IUPAC-namn | 2-[(E)-[2-[2-[2-[(E)-[1,3-dioxo-1-[(2-oxo-1,3-dihydrobenzimidazol-5-yl)amino]butan-2-yl]diazenyl]phenoxy]ethoxy]phenyl]diazenyl]-3-oxo-N-(2-oxo-1,3-dihydrobenzimidazol-5-yl)butanamide |
| Molekylvikt (g/mol) | 732.7 |
| Formel vikt | 732.2405 |