missing translation for 'onlineSavingsMsg'
Learn More

PIGS Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 18334-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16878414

  • 1211.96 SEK / 20µL
Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Component of the GPI transamidase complex. Essential for transfer of GPI to proteins, particularly for formation of carbonyl intermediates.
Specifications

Specifications

PIGS
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6PD26, Q96S52
PIGS
Rabbit
IgG
20 μL
-20°C
Primary
276846, 94005
Western Blot, Immunofluorescence, Immunocytochemistry
Unconjugated
PIGS
Liquid
PIGS
PIGS Fusion Protein Ag12950
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Videos
SDS
Documents
Certificates
Special Offers

Special Offers