Läs mer
PIK-75, TRC
Produktkod. 30412948
Change view
Kvantitet:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30412948 1 mg
30428222 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30412948 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP4376701MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 372196-67-3
Molekylformel C16H14BrN5O4S
Molekylvikt (g/mol) 452.28
Synonym 2-Methyl-5-nitro-benzenesulfonic Acid 2-[(6-Bromoimidazo[1,2-a]pyridin-3-yl)methylene]-1-methylhydrazide
IUPAC-namn N'-((6-bromoimidazo[1,2-a]pyridin-3-yl)methylene)-N,2-dimethyl-5-nitrobenzenesulfonohydrazide
LEDER O=N(C1=CC=C(C)C(S(=O)(N(C)N=CC2=CN=C3C=CC(Br)=CN32)=O)=C1)=O
Specifikationer
Kemiskt namn eller material 2-Methyl-5-nitro-benzenesulfonic Acid 2-[(6-Bromoimidazo[1,2-a]pyridin-3-yl)methylene]-1-methylhydrazide
Kvantitet 1 mg
InChI formel InChI=1S/C16H14BrN5O4S/c1-11-3-5-13(22(23)24)7-15(11)27(25,26)20(2)19-9-14-8-18-16-6-4-12(17)10-21(14)16/h3-10H,1-2H3
Formel vikt 450.99