missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIK3C2B (PI3K-C2 beta) Recombinant Human Protein
The class II PI3 kinases, including PIK3C2B, phosphorylate the 3 position of PI or PI4P.
18260.00 SEK - 79000.00 SEK
Specifications
Accession Number | NP_002637.2 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol |
Molecular Weight (g/mol) | 167.4kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15468703
|
Invitrogen™
PR9006A |
100μg |
18260.00 SEK
100µg |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15498783
|
Invitrogen™
PV5375 |
1mg |
79000.00 SEK
1mg |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Though not as well studied as the class I kinases, the class II PI3 enyzmes have been linked to diverse, recpetor-mediated cellular processes including insulin signalling, neuronal survival, and growth factor signalling.
Specifications
NP_002637.2 | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
PI3 Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
167.4kDa | |
RUO - research use only | |
PIK3C2B (PI3K-C2 beta) | |
Human | |
GST Tag | |
Frozen |