Läs mer
Invitrogen™ Human PIK3C2G (PI3K-C2 gamma), GST Tag Recombinant Protein
Beskrivning
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This PIK3C2G (PI3K-C2 gamma) recombinant human protein (468-1203) was expressed in insect cells. PIK3C2G (PI3K-C2 gamma) recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Adapta™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Recombinant, Active, Catalytic The gene accession number for PIK3C2G (PI3K-C2 gamma) is AAI30278.1.
Specifikationer
Specifikationer
| Tillträdesnummer | O75747 |
| Koncentration | See Label |
| För användning med (applikation) | Kinase Assay |
| Formulering | 50mM tris HCl with 0.1mM PMSF, 0.25mM DTT, 25% glycerol, 0.1mM EDTA, 10mM glutathione, 150mM NaCl and no preservative; pH 7.5 |
| Gen-ID (Entrez) | 5288 |
| Molekylvikt (g/mol) | 110 kDa |
| Namn | Human PIK3C2G (PI3K-C2 gamma), GST Tag |
| pH-intervall | 7.5 |
| Reningsmetod | Purified |
| Kvantitet | 100 μg |
| Visa mer |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.