missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIK3CG (p110 gamma) Recombinant Human Protein
PIK3CG (p110 gamma) is the sole class IB phosphatidylinositol 3-kinase family member.
17980.00 SEK - 138200.00 SEK
Specifications
Accession Number | NP_002640.2 |
---|---|
Formulation | 50mM Tris-HCl pH 7.5, 150mM NaCl, 0.1mM EGTA, 2mM DTT, 10% Glycerol |
Molecular Weight (g/mol) | 130.5kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Regulatory Status | RUO - research use only |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15438683
|
Invitrogen™
PR8641C |
100μg |
17980.00 SEK
100µg |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15448753
|
Invitrogen™
PV4787 |
1mg |
138200.00 SEK
1mg |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
PIK3CG (p110 gamma), along with the class IA family members, generates the important second messenger PIP2. However, unlike the class IA enzymes which signal downstream of receptor tyrosine kinases, PIK3CG primarily signals downstream of GPCRs.
Specifications
NP_002640.2 | |
130.5kDa | |
RUO - research use only | |
PIK3CG (p110 gamma), PI3 Kinase gamma | |
Human | |
His Tag | |
Frozen |
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.1mM EGTA, 2mM DTT, 10% Glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
PI3 Kinase Family | |
Recombinant | |
Baculovirus |