Läs mer
Eurofins DiscoverX Pim1 Protein, Active
Beskrivning
Protein serine/threonine kinase PIM1 (pim-1-oncogene) is a member of a small family of highly homologous kinases, including PIM1, PIM2, and PIM3, and all members are expressed in the thymus. Overexpression of each of these members can promote lymphomagenesis in mice. PIM1 associates with Myb and phosphorylates the Myb DNA binding domain, suggesting that it regulates Myb protein activity by direct phosphorylation. Recently four proto-oncogenes PIM1, cMYC, PAX-5 and RhoH/TTF were found hypermutated in diffuse large B cell lymphoma (DLBCL) with >50% of cases carrying at least two mutated genes.
GenBank M27903
Specifikationer
Specifikationer
| Formulering | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 0.1 mM EGTA, 150 mM NaCl, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molekylvikt (g/mol) | 62 kDa |
| Kvantitet | 10 μg |
| Art | Human |
| Rekombinant | Recombinant |
| Protein Tag | N-terminal GST |
| Uttryckssystem | Recombinant enzyme expressed in E. coli cells |
| Proteinform | Full-length |
| Renhet eller kvalitetsklass | ≥73% |
| Protein | Pim-1, active |
Fisher Scientific Encompass-programmet erbjuder artiklar som inte ingår i vår distributionsportfölj. Dessa produkter har vanligtvis inga bilder eller detaljerade beskrivningar. Vi är dock angelägna om att förbättra din shoppingupplevelse. Använd formuläret nedan för att ge feedback relaterad till innehållet på denna produkt.