missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIM1 Recombinant Human Protein
PIM1 Recombinant Human Protein
Brand: Invitrogen PV3503
Additional Details : Weight : 0.05250kg
Description
The proto-oncogene PIM1 is upregulated in prostate cancer.Specifications
NP_002639 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
40.7kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
PIM1 | |
PIM1 was pre-diluted in enzyme dilution buffer and assayed in 50mM Tris (pH 7.5), 10mM MgCl2, 2mM DTT, 0.025% Triton™ X-100, 200μM ATP, 100μM PIM1 peptide substrate (AKKRRLSA) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
PIM1 Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
0.2mg/mL | |
5292 | |
PIM1 Recombinant Human Protein | |
–80°C | |
1 Each | |
PIM1 Recombinant Human Protein | |
PIM | |
16500nmoles of phosphate transferred to PIM1 peptide substrate (AKKRRLSA) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 0.083μg/mL | |
His Tag | |
Kinase and inhibitors | |
85% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.