Learn More
Invitrogen™ PIM2 Recombinant Human Protein
PIM2 is a proto-oncogene.
5380.00 SEK - 79000.00 SEK
Specifications
Accession Number | NP_006866 |
---|---|
Formulation | 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 63.7kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Regulatory Status | RUO - research use only |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11523487
|
Invitrogen™
PV3649 |
10μg |
5380.00 SEK
10µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15488473
|
Invitrogen™
PR6347B |
100μg |
18240.00 SEK
100µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10766983
|
Invitrogen™
PV4036 |
1mg |
79000.00 SEK
1mg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
PIM2 phosphorylates BAD, which partially inhibits its apopotopic function.
Specifications
NP_006866 | |
63.7kDa | |
RUO - research use only | |
PIM Kinase Family | |
Recombinant | |
Baculovirus |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Pim2 | |
Human | |
GST Tag |