Läs mer
Pimavanserin N-Oxide, TRC
Produktkod. 30417369
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30417369 25 mg
30416793 5 mg
30422038 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30417369 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP44180525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1134815-71-6
Molekylformel C25H34FN3O3
Molekylvikt (g/mol) 443.55
Synonym N-[(4-Fluorophenyl)methyl]-N-(1-methyl-1-oxido-4-piperidinyl)-N'-[[4-(2-methylpropoxy)phenyl]methyl]urea
IUPAC-namn 1-[(4-fluorophenyl)methyl]-1-(1-methyl-1-oxidopiperidin-1-ium-4-yl)-3-[[4-(2-methylpropoxy)phenyl]methyl]urea
LEDER CC(C)COc1ccc(CNC(=O)N(Cc2ccc(F)cc2)C3CC[N+](C)([O-])CC3)cc1
Specifikationer
Kemiskt namn eller material Pimavanserin N-Oxide
Kvantitet 25 mg
InChI formel InChI=1S/C25H34FN3O3/c1-19(2)18-32-24-10-6-20(7-11-24)16-27-25(30)28(17-21-4-8-22(26)9-5-21)23-12-14-29(3,31)15-13-23/h4-11,19,23H,12-18H2,1-3H3,(H,27,30)
Formel vikt 443.2584