Läs mer
Pimelic Diphenylamide 106 (analog), MedChemExpress
Produktkod. 16467616
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16467616 100 mg
16457768 10 mg
16423737 50 mg
16400238 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16467616 Leverantör Medchem Express Leverantörsnummer HY19430
MedChemExpress Pimelic Diphenylamide 106 analog is an analog of Pimelic Diphenylamide 106 with unknown biological activity.
Pimelic Diphenylamide 106 analog is an analog of Pimelic Diphenylamide 106 with unknown biological activity.
Specifikationer
Kemiskt namn eller material Pimelic Diphenylamide 106 (analog)
Fysisk form Solid
CAS 2070015-24-4
Färg White
Kvantitet 100 mg
Anteckningar om renhetsgrad Research
Molekylformel C21H27N3O2
För användning med (applikation) Cancer-programmed cell death
Synonym RGFA-8 analog TC-H 106 analog
Löslighetsinformation DMSO : ≥ 30 mg/mL (84.88 mM)
LEDER O=C(NC1=CC=CC=C1N)CCCCCCC(NC2=CC=C(C)C=C2)=O
Molekylvikt (g/mol) 353.46
Formel vikt 353.46
Procent renhet 98.2%