missing translation for 'onlineSavingsMsg'
Läs mer

Pimobendan, MedChemExpress

Produktkod. p-200096088
Encompass_Preferred
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 16438418

Brand: Medchem Express HYB020410mg

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

MedChemExpress Pimobendan (UD-CG115) is a selective inhibitor of PDE3 with IC50 of 0.32 μM.

Pimobendan (UD-CG115) is a selective inhibitor of PDE3 with IC50 of 0.32 μM.

Specifications

Kemiskt namn eller material Pimobendan
CAS 74150-27-9
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C19H18N4O2
Kvantitet 10 mg
Synonym UD-CG115
Löslighetsinformation DMSO : 50 mg/mL (149.53 mM; Need ultrasonic)
LEDER O=C1CC(C)C(C2=CC=C3N=C(C4=CC=C(OC)C=C4)NC3=C2)=NN1
Molekylvikt (g/mol) 334.37
Formel vikt 334.37
Procent renhet 99.76%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.