Läs mer
Pimodivir, MedChemExpress
Produktkod. 16467837
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16467837 5 mg
16445118 10 mg
16443296 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16467837 Leverantör Medchem Express Leverantörsnummer HY12353A
MedChemExpress Pimodivir (VX-787) is an orally bioavailable inhibitor of influenza A virus polymerases through interaction with the viral PB2 subunit.
Pimodivir (VX-787) is an orally bioavailable inhibitor of influenza A virus polymerases through interaction with the viral PB2 subunit.
Specifikationer
Kemiskt namn eller material Pimodivir
Fysisk form Solid
CAS 1629869-44-8
Färg White
Kvantitet 5 mg
Anteckningar om renhetsgrad Research
Molekylformel C20H19F2N5O2
För användning med (applikation) COVID-19-anti-virus
Synonym VX-787
Löslighetsinformation DMSO : 100 mg/mL (250.38 mM; Need ultrasonic)
LEDER O=C([C@H]1C(CC2)CCC2[C@@H]1NC3=NC(C4=CNC5=NC=C(F)C=C54)=NC=C3F)O
Molekylvikt (g/mol) 399.39
Formel vikt 399.39
Procent renhet 98.17%