Läs mer
Pinacolyl Methylphosphonate, TRC
Produktkod. 30416791
Change view
Kvantitet:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30416791 1 g
30415675 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30416791 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP4573051G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Pinacolyl methylphosphonic acid
CAS 616-52-4
Molekylformel C7 H17 O3 P
Kvantitet 1 g
Synonym Methylphosphonic acid','mono-(1,2,2-trimethylpropyl) ester
InChI formel InChI=1S/C7H17O3P/c1-6(7(2,3)4)10-11(5,8)9/h6H,1-5H3,(H,8,9)
LEDER CC(OP(=O)(C)O)C(C)(C)C
IUPAC-namn 3,3-dimethylbutan-2-yloxy(methyl)phosphinic acid
Molekylvikt (g/mol) 180.1818
Formel vikt 180.0915