Läs mer
Pindolol, TRC
Produktkod. 30180418
Change view
Kvantitet:
5 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180418 50 mg
30179800 100 mg
30179808 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30180418 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP46800050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13523-86-9
Molekylformel C14 H20 N2 O2
Molekylvikt (g/mol) 248.32
Synonym 2-Propanol, 1-(indol-4-yloxy)-3-(isopropylamino)- (8CI), 1-(1H-Indol-4-yloxy)-3-[(1-methylethyl)amino]-2-propanol, (RS)-Pindolol, (±)-4-[2-Hydroxy-3-(isopropylamino)propoxy]indole, (±)-LB 46, (±)-Pindolol, 1-[(1-Methylethyl)amino]-3-(4-indolyloxy)-2-propanol, 4-(2-Hydroxy-3-isopropylaminopropoxy)indole, 4-(3-Isopropylamino-2-hydroxypropoxy)indole, Apo-pindol, Betapindol, Blocklin L, Calvisken, Carvisken, DL-4-[2-Hydroxy-3-(isopropylamino)propoxy]indole, DL-LB 46, DL-Pindolol, Decreten, Durapindol, Glauco-Visken, LB 46, LB 46 (pharmaceutical), N-[2-Hydroxy-3-(1H-indol-4-yloxy)propyl]-N-isopropylamine, Pectobloc, Pinbetol, Pindolol, Prinodolol, Pynastin, Visken, dl-Pindolol
IUPAC-namn 1-(1H-indol-4-yloxy)-3-(propan-2-ylamino)propan-2-ol
LEDER CC(C)NCC(O)COc1cccc2[nH]ccc12
Specifikationer
Kemiskt namn eller material Pindolol
Kvantitet 50 mg
InChI formel InChI=1S/C14H20N2O2/c1-10(2)16-8-11(17)9-18-14-5-3-4-13-12(14)6-7-15-13/h3-7,10-11,15-17H,8-9H2,1-2H3
Formel vikt 248.1525