Läs mer
PINK1 Mouse anti-Human, Mouse, Rat, Clone: 8E10.1D6, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 7 publications
Brand: Novus Biologicals NBP2-36488
Beskrivning
PINK1 Monoclonal specifically detects PINK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, SDS-Page, Knockdown Validated.Specifikationer
| PINK1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 2-4 ug/ml, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 20-50 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml, SDS-Page, Knockdown Validated | |
| Q9BXM7 | |
| PINK1 | |
| PINK1 antibody was developed using a synthetic peptide made to the human PINK1 protein sequence (between residues 100-250). [Swiss-Prot: Q9BXM7] | |
| Protein G purified | |
| RUO | |
| 65018 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| 8E10.1D6 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| BRPK, EC 2.7.11.1, FLJ27236, PARK6, Parkinson disease (autosomal recessive) 6, PINK1 monoclonal, protein kinase BRPK, PTEN Induced Kinase 1, PTEN induced putative kinase 1, PTEN-induced putative kinase protein 1, serine/threonine-protein kinase PINK1, mit | |
| Mouse | |
| 62.7 kDa | |
| 0.1 mg | |
| Primary | |
| Human PINK1 protein sequence (between residues 100-250), only reactive to isoform 1. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b κ |
Fisher Scientific Encompass-programmet erbjuder artiklar som inte ingår i vår distributionsportfölj. Dessa produkter har vanligtvis inga bilder eller detaljerade beskrivningar. Vi är dock angelägna om att förbättra din shoppingupplevelse. Använd formuläret nedan för att ge feedback relaterad till innehållet på denna produkt.
For Research Use Only