Läs mer
Pinometostat, MedChemExpress
Produktkod. 16489217
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16489217 5 mg
16486658 10 mg
16422747 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16489217 Leverantör Medchem Express Leverantörsnummer HY15593
MedChemExpress Pinometostat (EPZ-5676) is a potent DOT1L histone methyltransferase inhibitor with a Ki of 80 pM.
Pinometostat (EPZ-5676) is a potent DOT1L histone methyltransferase inhibitor with a Ki of 80 pM.
Specifikationer
Kemiskt namn eller material Pinometostat
CAS 1380288-87-8
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C30H42N8O3
Kvantitet 5 mg
Synonym EPZ-5676
Löslighetsinformation DMSO : 100 mg/mL (177.71 mM; Need ultrasonic) ∣Ethanol : 100 mg/mL (177.71 mM; Need ultrasonic)
LEDER NC1=NC=NC2=C1N=CN2[C@H]3[C@H](O)[C@H](O)[C@@H](CN([C@@H]4C[C@H](CCC5=NC6=C(C=CC(C(C)(C)C)=C6)N5)C4)C(C)C)O3
Molekylvikt (g/mol) 562.71
Formel vikt 562.71
Procent renhet 98.99%
Kvalitet Research
Fysisk form Powder
För användning med (applikation) Cancer-programmed cell death