Läs mer
Pinonic Acid, TRC
Produktkod. 30423072
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30423072 500 mg
1 options
Produktkod. 30423072 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP468550500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material cis-Pinonic acid
CAS 473-72-3
Molekylformel C10 H16 O3
Kvantitet 500 mg
Synonym (1R,3R)-rel-3-acetyl-2,2-dimethyl-cyclobutaneacetic acid, (±)-cis-Pinonic acid, DL-cis-Pinonic acid
InChI formel InChI=1S/C10H16O3/c1-6(11)8-4-7(5-9(12)13)10(8,2)3/h7-8H,4-5H2,1-3H3,(H,12,13)/t7-,8+/m1/s1
LEDER CC(=O)[C@@H]1C[C@H](CC(=O)O)C1(C)C
IUPAC-namn 2-[(1R,3R)-3-acetyl-2,2-dimethyl-cyclobutyl]acetic acid
Molekylvikt (g/mol) 184.23
Formel vikt 184.1099