Läs mer
Pioglitazone Hydrochloride, TRC
Produktkod. 30041281
Change view
Kvantitet:
10 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30041281 100 mg
18063430 1 g
18043430 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30041281 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP471000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 112529-15-4
Molekylformel C19 H20 N2 O3 S . Cl H
Molekylvikt (g/mol) 392.9
Synonym 2,4-Thiazolidinedione, 5-[[4-[2-(5-ethyl-2-pyridinyl)ethoxy]phenyl]methyl]-, monohydrochloride (9CI), 2,4-Thiazolidinedione, 5-[[4-[2-(5-ethyl-2-pyridinyl)ethoxy]phenyl]methyl]-, monohydrochloride, (±)-, 5-[4-[2-(5-Ethyl-2-pyridyl)ethoxy]benzyl]thiazolidine-2,4-dione hydrochloride, AD 4833, Actos, Glutazone, Pioglitazone hydrochloride, Piomed, Poze, U 72107A
IUPAC-namn 5-[[4-[2-(5-ethylpyridin-2-yl)ethoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione;hydrochloride
LEDER Cl.CCc1ccc(CCOc2ccc(CC3SC(=O)NC3=O)cc2)nc1
Specifikationer
Kemiskt namn eller material Pioglitazone Hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C19H20N2O3S.ClH/c1-2-13-3-6-15(20-12-13)9-10-24-16-7-4-14(5-8-16)11-17-18(22)21-19(23)25-17;/h3-8,12,17H,2,9-11H2,1H3,(H,21,22,23);1H
Formel vikt 392.0961
Procent renhet >95%