Läs mer
Pioglitazone N-Oxide, TRC
Produktkod. 30423014
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30423014 2.5 mg 2.5mg
30422278 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30423014 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP4710102.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 145350-09-0
Molekylformel C19 H20 N2 O4 S
Molekylvikt (g/mol) 372.44
Synonym 5-[[4-[2-(5-Ethyl-1-oxido-2-pyridinyl)ethoxy]phenyl]methyl]-2,4-thiazolidinedione, 2,4-Thiazolidinedione, 5-[[4-[2-(5-ethyl-2-pyridinyl)ethoxy]phenyl]methyl]-, N-oxide, Pioglitazone N-oxide
IUPAC-namn 5-[[4-[2-(5-ethyl-1-oxidopyridin-1-ium-2-yl)ethoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione
LEDER CCc1ccc(CCOc2ccc(CC3SC(=O)NC3=O)cc2)[n+]([O-])c1
Specifikationer
Kemiskt namn eller material Pioglitazone N-Oxide
Kvantitet 2.5 mg
InChI formel InChI=1S/C19H20N2O4S/c1-2-13-3-6-15(21(24)12-13)9-10-25-16-7-4-14(5-8-16)11-17-18(22)20-19(23)26-17/h3-8,12,17H,2,9-11H2,1H3,(H,20,22,23)
Formel vikt 372.1144