Läs mer
Pipazetate, TRC
Produktkod. 30417525
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30417525 10 mg
1 options
Produktkod. 30417525 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP47570510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Pipazetate
CAS 2167-85-3
Molekylformel C21 H25 N3 O3 S
Kvantitet 10 mg
Synonym 10H-Pyrido[3,2-b][1,4]benzothiazine-10-carboxylic acid','2-[2-(1-piperidinyl)ethoxy]ethyl ester, 10H-Pyrido[3,2-b][1,4]benzothiazine-10-carboxylic acid','2-(2-piperidinoethoxy)ethyl ester (7CI,8CI), Ethanol','2-(2-piperidinoethoxy)-','10H-pyrido[3,2-b][1,4]benzothiazine-10-carboxylate (ester) (8CI), 2-(2-Piperidinoethoxy)ethyl 1-azaphenothiazine-10-carboxylate, 2-(2-Piperidinoethoxy)ethyl 10H-pyrido[3,2-b][1,4]benzothiazine-10-carboxylate, D 254, Pipazetate, Pipazethate, SKF 70230A, SQ 15874
InChI formel InChI=1S/C21H25N3O3S/c25-21(27-16-15-26-14-13-23-11-4-1-5-12-23)24-17-7-2-3-8-18(17)28-19-9-6-10-22-20(19)24/h2-3,6-10H,1,4-5,11-16H2
LEDER O=C(OCCOCCN1CCCCC1)N2c3ccccc3Sc4cccnc24
IUPAC-namn 2-(2-piperidin-1-ylethoxy)ethyl pyrido[3,2-b][1,4]benzothiazine-10-carboxylate
Molekylvikt (g/mol) 399.51
Formel vikt 399.1617