Läs mer
Pipendoxifene, TRC
Produktkod. 30422959
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30422959 100 mg
30424372 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30422959 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP479850100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 198480-55-6
Molekylformel C29 H32 N2 O3
Molekylvikt (g/mol) 456.58
Synonym 2-(4-hydroxyphenyl)-3-methyl-1-[[4-[2-(1-piperidinyl)ethoxy]phenyl]methyl]-1H-indol-5-ol, ERA 923
IUPAC-namn 2-(4-hydroxyphenyl)-3-methyl-1-[[4-(2-piperidin-1-ylethoxy)phenyl]methyl]indol-5-ol
LEDER Cc1c(c2ccc(O)cc2)n(Cc3ccc(OCCN4CCCCC4)cc3)c5ccc(O)cc15
Specifikationer
Kemiskt namn eller material Pipendoxifene
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C29H32N2O3/c1-21-27-19-25(33)11-14-28(27)31(29(21)23-7-9-24(32)10-8-23)20-22-5-12-26(13-6-22)34-18-17-30-15-3-2-4-16-30/h5-14,19,32-33H,2-4,15-18,20H2,1H3
Formel vikt 456.2413
Procent renhet >95%